Learn More
Alkali Scientific ASI™ 96-Well Hanging Drop Plate, 123.6 x 81.4 x 9mm, 30μl Droplet Volume, PS, Sterile, Without 96 Well Cell Culture Plate, 10/Cs
SDP
Catalog No. 503027033
Encompass_Preferred
Our 96 Well Hanging Drop Plate are suitable for spheroid culture. The shape of the holes is designed to position droplets safely. This shape keeps the droplets of the hanging drop consistent and stable culturing conditions. The reservoir prevents evaporation of the media, allowing for long-term culture. The tube extending into the well plate facilitates collect of spheroids. • Physiological and non-expensive spheroid culture system• Suitable for long-term culture ▪ Non – pyrogenic▪ DNase / RNase – free ▪ Human DNA – free ▪ Non – cytotoxic • Suitable for high-throughput screening (HTS)
Catalog No. 50-302-7033 Supplier Alkali Scientific Supplier No. HDP0096
Add to Cart
Add to Cart
Our 96 Well Hanging Drop Plate are suitable for spheroid culture. The shape of the holes is designed to position droplets safely. This shape keeps the droplets of the hanging drop consistent and stable culturing conditions. The reservoir prevents evaporation of the media, allowing for long-term culture. The tube extending into the well plate facilitates collect of spheroids. • Physiological and non-expensive spheroid culture system• Suitable for long-term culture ▪ Non – pyrogenic▪ DNase / RNase – free ▪ Human DNA – free ▪ Non – cytotoxic • Suitable for high-throughput screening (HTS)