Learn More
Molecular Devices Molecular Devices Cell Wash Head, 96-Well, Angled Dispense Probes, Interchangeable
Catalog No. 5000014687
Encompass_Preferred
The Molecular Devices AquaMax 96-Well Cell Wash Head is a replacement cell wash head engineered for gentle, controlled washing of cell-based microplate assays. It is designed for use with AquaMax series microplate washers and is interchangeable with other wash head configurations.
  • Configuration: 96-well cell wash head.
  • Angled dispense probes minimize cell loss during washing.
  • Interchangeable with other wash heads and detected by instrument software.
  • Removable lever mounting for easy installation and removal.
  • Clear probe guard protects probes and reduces spills and sprays.
Catalog No. 50-000-14687 Supplier Molecular Devices Supplier No. 03105217T
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices AquaMax 96-Well Cell Wash Head is a replacement cell wash head engineered for gentle, controlled washing of cell-based microplate assays. It is designed for use with AquaMax series microplate washers and is interchangeable with other wash head configurations.
  • Configuration: 96-well cell wash head.
  • Angled dispense probes minimize cell loss during washing.
  • Interchangeable with other wash heads and detected by instrument software.
  • Removable lever mounting for easy installation and removal.
  • Clear probe guard protects probes and reduces spills and sprays.