Learn More
Greiner Bio-One 96-well High Binding Standard ELISA Microplates
GREENER_CHOICE
Catalog No. 07000101 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Square, Flat Bottom (F-well)
Square, Round Bottom (U-well)
Sterility:
Non-sterile
Sterile
Well Design:
Chimney
Round
Standard
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Sterility Well Design
07-000-101 Flat Bottom (F-well) Non-sterile Chimney
07-000-102 Square, Flat Bottom (F-well) Non-sterile Standard
07-000-608 Square, Round Bottom (U-well) Non-sterile Round
07-000-995 Round Bottom (U-well) Sterile Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-101 Supplier Greiner Bio-One Supplier No. 655081
Add to Cart
Edge
Add to Cart
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9mm
  • Specifically tre
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon 600
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Chimney