Learn More
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9 mm
Service Plan
No
Certifications
DIN ISO 9001
Warranty and Services
No
Specifications
Specifications
| Barcode | Alphanumeric |
| Format | Standard |
| Array | 8 x 12 |
| Binding Type | High Binding |
| Color | Clear |
| Product Type | ELISA Microplate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Microlon 600 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.