Learn More
Genscript Corporation 96-well Low Binding White Micr
Catalog No. 5000058353
Encompass
The 96-well low-binding white microplates are applicable for ELISA assay These microplates have a corner notch for use with standard plate readers
Catalog No. 50-000-58353 Supplier Genscript Corporation Supplier No. D00042
The 96-well low-binding white microplates are applicable for ELISA assay These microplates have a corner notch for use with standard plate readers