Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural
GSA_VA

Catalog No. 14230239 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-230-239 Blue
14-230-241 Yellow
14-230-240 Green
14-230-237 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-230-239
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Fit most thermal cyclers

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Stackable
  • Raised-rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 200μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • SBS (Society for Biomolecular Screening) footprint

  • Automation friendly
  • Eight holes in the skirt aid plate positioning and removal from thermal cycler block
  • Cleanroom produced

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Blue
Product Type PCR Microplate
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes
Well Color Clear
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.