Learn More
Microsolv Technology Corporation 96 Well Mat Square Preslit
Catalog No. 5001452278
Encompass_Preferred
Sealing Mat, Pre-Slit Silicone Rubber, for 96 Well Plates, Square Wells. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52278 Supplier Microsolv Technology Corporation Supplier No. 9509658
Sealing Mat, Pre-Slit Silicone Rubber, for 96 Well Plates, Square Wells. MicroSolv Brand. 1/EA.