Learn More
Greiner Bio-One 96-well Medium Binding Standard ELISA Microplates
GREENER_CHOICE
Catalog No. 07000613 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
07-000-613 Conical Bottom (V-well)
07-000-100 Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-613 Supplier Greiner Bio-One Supplier No. 651001
Add to Cart
Add to Cart
For diagnostics and immunological research applications. Greiner Bio-One 96-Well Medium Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9 mm
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Array 8 x 12
Packaging Inner bags of 10
Binding Type Medium Binding
Color Clear
Product Type ELISA Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Medium Binding
Well Shape Conical Bottom (V-well)
Quantity 40/Cs.