Learn More
Greiner Bio-One 96-well MICROLON™ 600 Flat Bottomed Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07001000 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-001-000 Black
1 options
Catalog No. 07-001-000 Supplier Greiner Bio-One Supplier No. 65506125
Add to Cart
Add to Cart
Ideal for work with nonpolar proteins and peptides
  • Features high binding
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Binding Type High Binding
Color Black
Material Polystyrene
No. of Wells 96
Well Shape Flat Bottom (F-well)
Well Design Chimney