Learn More
Quality Biological Inc Corning 96-well Clear Flat Bottom Polystyrene High Bind Microplate, 25 per Bag, without Lid, Nonsterile
SDP
Catalog No. NC2055453
Encompass_Preferred
Ideal for immunoassays. High binding surface is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
• Binding capacity is approximately 400 to 500ng IgG/cm²
• Without lids, lids supplied separately
• Total volume is 360 µL; recommended working volume of 75 to 200 µL
• Conforms to standard microplate footprint and dimensions
• Cell culture microplates are sterile and nonpyrogenic.
• Lids available where indicated
Catalog No. NC2055453 Supplier Quality Biological Inc Supplier No. P6589018
Ideal for immunoassays. High binding surface is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
• Binding capacity is approximately 400 to 500ng IgG/cm²
• Without lids, lids supplied separately
• Total volume is 360 µL; recommended working volume of 75 to 200 µL
• Conforms to standard microplate footprint and dimensions
• Cell culture microplates are sterile and nonpyrogenic.
• Lids available where indicated