Learn More
Thomas Scientific 96-well microplate, round u-bottom, untreated, non-sterile, polystyrene
SDP
Catalog No. 5001112
Encompass
96-well microplates are available in flat (F), conical (V), and round (U) bottom varieties. Polypropylene microplates offer higher temperature resistance and are less reactive than polystyrene. Each well in the microplate is designed to have a raised rim to reduce cross contamination, and to provide a better fit to the corresponding raised ring in the lid. Lid use reduces excessive evaporative fluid loss. Sterile plates are gamma irradiated.
Catalog No. 50-011-12 Supplier Thomas Scientific Supplier No. 1185U55
96-well microplates are available in flat (F), conical (V), and round (U) bottom varieties. Polypropylene microplates offer higher temperature resistance and are less reactive than polystyrene. Each well in the microplate is designed to have a raised rim to reduce cross contamination, and to provide a better fit to the corresponding raised ring in the lid. Lid use reduces excessive evaporative fluid loss. Sterile plates are gamma irradiated.