Learn More
Greiner Bio-One 96-well No Bottom Microplates
GREENER_CHOICE
Catalog No. 07000626 Shop All Greiner Bio-One Products
Change view
Click to view available options
No. of Wells:
96
Catalog No. No. of Wells
07-000-626 96
1 options
Catalog No. 07-000-626 Supplier Greiner Bio-One Supplier No. 655000
Add to Cart
Add to Cart
Made of polystyrene. Greiner Bio-One 96-Well No Bottom Microplates are designed without bottoms.
  • Virgin polystyrene construction
  • Chimney-well design to help prevent cross-contamination
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Array 8 x 12
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape No Bottom
Quantity 40/Cs.
Well Design Chimney