Learn More
Bellco Glass, Inc. 96 Well Non-treated Plate with
SDP
Catalog No. 502330554
Encompass_Preferred
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume50-300 μLBottom ShapeFlatTissue Culture TreatedNoRNase/DNaseYes
Catalog No. 50-233-0554 Supplier Bellco Glass, Inc. Supplier No. 229596000
Add to Cart
Add to Cart
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume50-300 μLBottom ShapeFlatTissue Culture TreatedNoRNase/DNaseYes