Learn More
Bellco Glass, Inc. 96 Well Non-Treated Plate with
SDP
Catalog No. 502329571
Encompass_Preferred
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates96 Well Non-Treated Plate now available in 5/Pack for large-scale users Pack Description5/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume50-300 μLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes
Catalog No. 50-232-9571 Supplier Bellco Glass, Inc. Supplier No. 229597000
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates96 Well Non-Treated Plate now available in 5/Pack for large-scale users Pack Description5/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume50-300 μLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes