Learn More
Greiner Bio-One 96-well Non-treated Polystyrene Microplates
Catalog No. 07000609 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Sterility:
Non-sterile
Sterile
Quantity:
100/Cs.
375/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Sterility Quantity
07-000-609 Round Bottom (U-well) Non-sterile 100/Cs.
07-000-124 Flat Bottom (F-well) Non-sterile 100/Cs.
07-000-140 Conical Bottom (V-well) Non-sterile 100/Cs.
07-000-610 Round Bottom (U-well) Sterile 375/Cs.
07-000-611 Round Bottom (U-well) Non-sterile 375/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-000-609 Supplier Greiner Bio-One Supplier No. 650101
Add to Cart
Edge
Add to Cart
For agglutination tests. Greiner Bio-One 96-Well Non-treated Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Polystyrene
  • Can be traced all the way back to production through a defined LOT number system
  • No sharp corners, can be pipetted easily and cleanly
    Service Plan
    No
    Certifications
    DIN ISO 9001
    Warranty and Services
    No
    TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
No. per Case 100
Quantity 100/Cs.