Learn More
Electron Microscopy Sciences NUNC™ 96 Well Bottom Plates Glass, Black, Non-treated, Sterile
SDP
Catalog No. 5033300
Encompass_Preferred
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm. Black upper structure with #1.5 borosilicate coverglass. Non-treated plates are optimized for fluorescence.
Color: Black
Type: Non-treated
Sterile: Sterile
Catalog No. 50-333-00 Supplier Electron Microscopy Sciences Supplier No. 6481515
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm. Black upper structure with #1.5 borosilicate coverglass. Non-treated plates are optimized for fluorescence.
Color: Black
Type: Non-treated
Sterile: Sterile