Learn More
Electron Microscopy Sciences NUNC™ 96 Well Bottom Plates Black, CC2, Sterile
SDP
Catalog No. 5033299
Encompass_Preferred
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm. Black upper structure with #1.5 borosilicate coverglass. Sterile with lid. CC2 surface is superior for attachment and mimics Poly-D-Lysine.
Color: Black
Type: CC2
Sterile: Sterile
Catalog No. 50-332-99 Supplier Electron Microscopy Sciences Supplier No. 6481520
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm. Black upper structure with #1.5 borosilicate coverglass. Sterile with lid. CC2 surface is superior for attachment and mimics Poly-D-Lysine.
Color: Black
Type: CC2
Sterile: Sterile