Learn More
Electron Microscopy Sciences NUNC™ 96 Well Bottom Plate White, Cell Culture, Sterile
SDP
Catalog No. 5033298
Encompass_Preferred
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm. White upper structure with #1.5 borosilicate coverglass. Sterile with lid.
Color: White
Type: Cell Culture
Sterile: Sterile
Catalog No. 50-332-98 Supplier Electron Microscopy Sciences Supplier No. 6481510
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm. White upper structure with #1.5 borosilicate coverglass. Sterile with lid.
Color: White
Type: Cell Culture
Sterile: Sterile