Learn More
Electron Microscopy Sciences NUNC™ 96 Well Optical Bottom Plate Black, Cell Culture Sterile
SDP
Catalog No. 5030876
Encompass_Preferred
The NUNC Coverglass Optical Bottom 96 Well Plates have a polymer bottom and combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. The optical clarity allows for easy viewing of well contents. Footprint compatible with standard equipment and automated systems. It has a flat bottom well geometry for plate reader access and working ranges from 50-200 uL/well. Black upper structure with Polymer bottom
Color: Black
Surface: Cell Culture
Sterile: Sterile
This item is not returnable. View return policy
Catalog No. 50-308-76 Supplier Electron Microscopy Sciences Supplier No. 6481015
The NUNC Coverglass Optical Bottom 96 Well Plates have a polymer bottom and combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. The optical clarity allows for easy viewing of well contents. Footprint compatible with standard equipment and automated systems. It has a flat bottom well geometry for plate reader access and working ranges from 50-200 uL/well. Black upper structure with Polymer bottom
Color: Black
Surface: Cell Culture
Sterile: Sterile