Learn More
Electron Microscopy Sciences NUNC™ 96 Well Optical Bottom Plate Black, Non-treated
SDP
Catalog No. 5030880
Encompass_Preferred
The NUNC Coverglass Optical Bottom 96 Well Plates have a polymer bottom and combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. The optical clarity allows for easy viewing of well contents. Footprint compatible with standard equipment and automated systems. It has a flat bottom well geometry for plate reader access and working ranges from 50-200 uL/well. Non-treated surface is optimal for scintillation counting. Black
Color: Black
Surface: Non-treated
Catalog No. 50-308-80 Supplier Electron Microscopy Sciences Supplier No. 6481110
The NUNC Coverglass Optical Bottom 96 Well Plates have a polymer bottom and combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. The optical clarity allows for easy viewing of well contents. Footprint compatible with standard equipment and automated systems. It has a flat bottom well geometry for plate reader access and working ranges from 50-200 uL/well. Non-treated surface is optimal for scintillation counting. Black
Color: Black
Surface: Non-treated