Learn More
Zymo Research Corporation 96-Well PCR Conversion Plate w/ Cover Foil (2 Plates/Foils)
SDP
Catalog No. 502037013
Encompass_Preferred
The plate can accommodate up to 150 l, suitable for bisulfite conversion processes. Includes a cover foil that can be pierced for easy downstream processing.
Catalog No. 50-203-7013 Supplier Zymo Research Corporation Supplier No. C2005
The plate can accommodate up to 150 l, suitable for bisulfite conversion processes. Includes a cover foil that can be pierced for easy downstream processing.