Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ 96-Well Microplates, BIO-CERT™ PCR QUALITY, Standard Profile and Elevated Rim

Catalog No. 1388275 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Skirt Style:
Non-skirt
Semi-skirted
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Skirt Style
13-882-75 Semi-skirted
13-882-74 Non-skirt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-882-75 Supplier BRANDTECH™ BRAND™ Supplier No. 781400
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for reliable and precise results in a wide range of PCR applications.

BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Versatile Formats
    • Suitable for most commercially available thermal cyclers
    • Non-skirted plates are highly flexible and compatible with most thermal cyclers
    • Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color Clear
Disposable Single-use
Product Type PCR 96-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Semi-skirted
Material Polypropylene
Volume (Metric) Well 0.20 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
Includes Plates only
No. per Pack 50/Pk.
Profile Standard
RNase Free Yes
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.