Learn More
PERMAGEN LABWARE LTD 96-Well PCR Post Magnet Low Elution Plate
Catalog No. NC1880783
Encompass_Preferred
The Permagen post magnet low elution plate was designed for use in automation applications where volumes as low as 5 uL are required. Most PCR plates are bent, leading to inconsistent lab results. This plate pulls the magnetic beads to the side of each well.
Catalog No. NC1880783 Supplier PERMAGEN LABWARE LTD Supplier No. LE400
Add to Cart
Add to Cart
The Permagen post magnet low elution plate was designed for use in automation applications where volumes as low as 5 uL are required. Most PCR plates are bent, leading to inconsistent lab results. This plate pulls the magnetic beads to the side of each well.