Learn More
Research Products International Corp 96-Well Plate Cover Foil, Pierceable, 24 Foils
SDP
Catalog No. 501978237
Encompass
Pierceable aluminum foil with strong adhesive strength for sealing 96-well plates and blocks. Ideal for cold storage. Dimensions are 82.6 mm x 132.6 mm.
Catalog No. 50-197-8237 Supplier Research Products International Corp Supplier No. ZC200724
Pierceable aluminum foil with strong adhesive strength for sealing 96-well plates and blocks. Ideal for cold storage. Dimensions are 82.6 mm x 132.6 mm.