Learn More
Electron Microscopy Sciences MicroWell 96-Well Plate, Flat with Lid, 0.4ml, 0.328cm2 Culture Area
SDP
Catalog No. 501899309
Encompass_Preferred
MicroWell Plates are used in a broad spectrum of applications, including cloning experiments, sample incubations, and in tissue culture based analytical systems. Designed to be stackable, are radiation sterilized, and have excellent cell attachment.
Type: Flat with Lid
Volume: 0.4ml
Culture Area: 0.328cm2
Catalog No. 50-189-9309 Supplier Electron Microscopy Sciences Supplier No. 70437F5
Add to Cart
Add to Cart
MicroWell Plates are used in a broad spectrum of applications, including cloning experiments, sample incubations, and in tissue culture based analytical systems. Designed to be stackable, are radiation sterilized, and have excellent cell attachment.
Type: Flat with Lid
Volume: 0.4ml
Culture Area: 0.328cm2