Learn More
LICORbio 96-Well Plate for In-Cell Western™ Assays (8 plates)
Catalog No. NC2002726
Encompass_Preferred
Sterile cell culture plates optimized for use for the In-Cell Western Assay. These plates include a lid and have a clear, polystyrene bottom with black walls.
Catalog No. NC2002726 Supplier LICORbio Supplier No. 92619156
Sterile cell culture plates optimized for use for the In-Cell Western Assay. These plates include a lid and have a clear, polystyrene bottom with black walls.