Learn More
Waters Corp WATERS CORPORATION 96-well Plate, Polypropylene, for Glass Inserts, 20/pk
Catalog No. 50780698 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well Plate, made of polypropylene, prevents chemical reactions. Designed for autosamplers, it is SBS/ASI compliant for robot-compatible systems and serves as a collecting plate for SPE and filtration layouts. Compatible with glass inserts, it protects against contaminants, temperature changes, and organic solvents when used with a sealant. It supports ACQUITY UPLC Technology for enhanced resolution, sensitivity, and throughput.
  • Made of polypropylene
  • Designed for autosamplers and robot-compatible systems (SBS/ASI compliant)
  • Functions as collecting plates for 96-well SPE and filtration layouts
  • Compatible with glass inserts
  • Provides protection against contaminants, temperature changes, and organic solvents (with sealant)
  • Centrifugable up to 5000g
  • Supports ACQUITY UPLC Technology
  • Compatible with ACQUITY UPLC and ACQUITY UPLC I-Class FL systems
Catalog No. 50-780-698 Supplier Waters Corp Supplier No. 186001438
This 96-well Plate, made of polypropylene, prevents chemical reactions. Designed for autosamplers, it is SBS/ASI compliant for robot-compatible systems and serves as a collecting plate for SPE and filtration layouts. Compatible with glass inserts, it protects against contaminants, temperature changes, and organic solvents when used with a sealant. It supports ACQUITY UPLC Technology for enhanced resolution, sensitivity, and throughput.
  • Made of polypropylene
  • Designed for autosamplers and robot-compatible systems (SBS/ASI compliant)
  • Functions as collecting plates for 96-well SPE and filtration layouts
  • Compatible with glass inserts
  • Provides protection against contaminants, temperature changes, and organic solvents (with sealant)
  • Centrifugable up to 5000g
  • Supports ACQUITY UPLC Technology
  • Compatible with ACQUITY UPLC and ACQUITY UPLC I-Class FL systems