Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 96-well Plate Frame for 1x12 Strip Wells

Catalog No. 07000774 Shop All Greiner Bio-One Products
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07-000-774 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-774 Supplier Greiner Bio-One Supplier No. 701185

For use with 96-well microplate. Greiner Bio-One 96-Well Plate Frame for 1x12 Strip Wells holds microplate in place.

  • 96W Plate Frame for F12 Strips (1x12 Strip Wells)
TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Color White
Material Polystyrene
Sterility Non-sterile
Quantity 400/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.