Learn More
Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats
Shop All Thermo Scientific Products

Description
Save time and minimize the risk of cross-contamination with convenient 96-Well Plate Kits packaged with pre-assembled glass or PTFE vials.
Save time and minimize the risk of cross-contamination with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates, pre-inserted glass or PTFE vials and silicone/PTFE sealing mats.
- Convenient wellplate kits include pre-inserted glass or PTFE vials and 96-position sealing mats; some include cutting tool
- Mats seal samples for storage
- Plates are glass-inserted for greater inertness compared to pure PP plates
- Unique cutting tool allows removal of individual samples with the seal in place, eliminating cross-contamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Packaging | 5 Pk. |
| Product Line | Webseal |
| Color | Clear |
| Product Type | Glass Inserted Plate Kit with 96-Position Mat |
| Material | Silicone, PTFE, Glass |
| No. of Wells | 96 |
| Total Volume (Metric) Well | 1100 μL |
| Well Shape | Round |
| Show More |
Frequently Asked Questions (FAQs)
Yes, bar-coded WebSeal plates are available as a catalog product for the famly of Thermo Scientific Vanquish UHPLC machines only. However, plates can be provided with a pre-printed adhesive label applied. Contact your sales representative to assist with completing a form where you can define the bar code information and location of application onto the plate. Additional information required includes the physical requirements for the label such as temperature stability, solvent stability, etc.
WebSeal Plate+ or plates with glass or PTFE inserts should never be sealed with tapes or adhesive foils. Silicone mats should be used with the WebSeal Plate&plus products, preferably with a protective PTFE film to give increased chemical protection.
The glass inserted plates are sealed with specific silicone plug mats or individual caps. Again, most have additional PTFE films of layers to give increased chemical protection.
These plates, inserts, and mats were originally designed for combinatorial chemistry techniques with in situ chemical derivatization. Use with elevated temperatures is possible with the inserts and plugs. These are the closest plate format equivalent to glass autosampler vial products.
The WebSeal Plate+ coating provides additional surface protection that further reduces the possibility of interactions with solvents, sample component, or other materials. WebSeal Plate+ microptates display ultra-low background extractable organics profiles that compare favorably to our mass spectrometry certified vial products.
Well plates are manufactured in molds. In order to ''force'' the ready produced plate to come out of this mold for the next shot, some manufacturers use release agents (organic compounds that act as lubricants). These release agents can be detected in your chromatogram. The grade of polypropylene used by most plate manufacturers can contain monomers or other added ingredients that are susceptible to leaching on contact with organic solvents. WebSeal-Certified products are manufactured from high purity resins and use a process that does not require mold release agents and that is why they exhbit no significant background extractables.
If you are using high concentrations of organic solvents in your sample, then you may be concerned about ''bleed''. This is a term which normally implies a constant loss of material from the column or separation system which leads to high background levels, increased noise, and loss of sensitivity. If using solutions with a high level of buffer or water, then this should not be a concern as such components will not leach under these conditions. All plates may be used.
The evaluation of the plastic resins used in the production of plates was also carried out by controlled extraction of leachable components with organic solvents associated with HPLC and GC operation. These included methanol, acetonitrile, and hexane. Standard plates showed low level of extractables when tested at the highest instrument sensitivity range. These extractables found are characteristic of the common materials used for microplates in the clinical market and plates that have been used for general purpose chromatography applications. This indicates that for many non-critical applications and with higher concentrations of sample, the standard quality plates will be acceptable. These plates are termed Chromatography Tested.
The very low levels of background extractables found during our testing of some plates showed for applications run at more sensitive instrument settings. Where higher purity of the plate is required, you should directly switch to the Certified plates as they are the lowest extraction PP plates currently available on the world market. For critical analysis with high organic solvent concentrations and low sample concentrations, the Certified product range should be the plates of choice.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.