Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ HT 100 Plate Handler, Manual Track Switching

Catalog No. BTX450400 Shop All BTX Harvard Products
Change view
Click to view available options
Connection Type:
Banana Cables
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX450400 Banana Cables
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX450400 Supplier BTX™ Supplier No. 450400

missing translation for 'validMainframeMsg'

Designed to work with 96 well plates. BTX™ Harvard Apparatus 96 Well Plate Handler helps to create a safe and reliable working area for processing electroporation plates.

  • Quickly optimize for better gene delivery
  • Designed to work with 96 well plates
  • Pulse switching technology integrated into package and gold plated contacts to mate with electroporation plate
  • With an adapter to accommodate 25 well plates as well with our ECM 630 generator
  • Prosess up to 96 wells in seconds

Specifications

Format 25-well, 96-well
For Use With (Application) High-Throughput Electroporation
For Use With (Equipment) ECM630, ECM830, Gemini X2 electroporation systems
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 3000 VDC, 250 VAC
Description HT-100 Microplate Handler
Display LCD
Electrode Shape Pin
Includes Manual track switching, Connectors for standard pulse generators
Pulse Number Range 1 to 99
Pulse Length Range 10 μs to 10 s
Operating Temperature (Metric) 5°C to 40°C
Quantity 1 Ea.
Relative Humidity 20 to 80%
Size (Metric) Gap 2 mm, 4 mm
Connection Type Banana Cables
Interface Digital User
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.