Learn More
Applied Biosystems™ 96-Well Plate Retainer & Base Set, for SeqStudio™ Flex
Catalog No. A49316
Encompass_Preferred
Change view
Click to view available options
Plate Compatibility:
Standard 96-well plates
Catalog No. Plate Compatibility
A49316 Standard 96-well plates
1 options
This item is not returnable. View return policy
Catalog No. A49316 Supplier Applied Biosystems™ Supplier No. A49316
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
This 96-well plate Retainer and Base Set is for use with 96-well sample plates on SeqStudio Flex Series genetic analyzers.
This 96-well plate Retainer and Base Set is for use with standard 96-well plates on SeqStudio Flex Series genetic analyzers.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SeqStudio™ Flex Series Genetic Analyzers
For Use With (Application) Sanger Sequencing and Fragment Analysis
Plate Compatibility Standard 96-well plates
Quantity 4 sets
When shouldn't I use the 3500 plate retainers on the SeqStudio Flex Series Genetic Analyzers?
If the automatic barcode workflow is enabled on the SeqStudio Flex Series Genetic Analyzer, using the 3500 plate retainers will cause a barcode error in the home screen.
What is different about the SeqStudio Flex Series Genetic Analyzer plate retainers compared with the 3500 plate retainers?
The SeqStudio Flex Series Genetic Analyzer plate retainers have one opening on the side to allow barcode reading while the 3500 plate retainers have two openings on the side which can cover the plate barcode.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov