Learn More
Electron Microscopy Sciences MicroWell 96-Well Plate, Round w/o Lid, 0.3ml, 0.36cm2 Culture Area
SDP
Catalog No. 5027291
Encompass
MicroWell Plates are used in a broad spectrum of applications, including cloning experiments, sample incubations, and in tissue culture based analytical systems. Designed to be stackable, are radiation sterilized, and have excellent cell attachment.
Type: Round w/o Lid
Volume: 0.3ml
Culture Area: 0.36cm2
Catalog No. 50-272-91 Supplier Electron Microscopy Sciences Supplier No. 70436R5
Add to Cart
Add to Cart
MicroWell Plates are used in a broad spectrum of applications, including cloning experiments, sample incubations, and in tissue culture based analytical systems. Designed to be stackable, are radiation sterilized, and have excellent cell attachment.
Type: Round w/o Lid
Volume: 0.3ml
Culture Area: 0.36cm2