Learn More
Waters Corp 96-well Collection Plate with 700 µL Glass Insert, 1/pk
Catalog No. 50819163 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
96-well sample collection plate, fitted with a 700 µL glass insert, for use in autosamplers. The glass insert maintains sample in contact only with a glass surface. This plate is SBS/ANSI compliant for robot compatible systems. This plate can also serve as a collection plate for 96-well SPE and filtration-plate formats. The collection plate is made of polypropylene, for chemical resistance.
Catalog No. 50-819-163 Supplier Waters Corp Supplier No. 186000349
96-well sample collection plate, fitted with a 700 µL glass insert, for use in autosamplers. The glass insert maintains sample in contact only with a glass surface. This plate is SBS/ANSI compliant for robot compatible systems. This plate can also serve as a collection plate for 96-well SPE and filtration-plate formats. The collection plate is made of polypropylene, for chemical resistance.