Learn More
Thomson Instrument Company 96-Well Plate, 2mL, Square Well, Pyramid Bottom, Indiv. Wrapped w/ Lid, Sterile
SDP
Catalog No. 501489304
Encompass_Preferred
Thomson Well Plates come in 6-, 24-, and 96-well formats with multiple well shapes and bottoms for screening, sample prep, or concentration. They’re ANSI/SLAS compatible and offered with matching seals and lids for clean, repeatable workflows and automation.
•Format & geometry options — square or round wells; pyramid, round, or V-bottoms to match your assay and culture needs.
•Automation-friendly fixed plate orientation for liquid handlers.
•Sealing ecosystem — capmats, gas-permeable airporous seals, foil seals, and lids.
•Collection-plate role — used as the receiving plate beneath filter plates in clarification workflows.
• Made in the USA
Catalog No. 50-148-9304 Supplier Thomson Instrument Company Supplier No. 931137
Thomson Well Plates come in 6-, 24-, and 96-well formats with multiple well shapes and bottoms for screening, sample prep, or concentration. They’re ANSI/SLAS compatible and offered with matching seals and lids for clean, repeatable workflows and automation.
•Format & geometry options — square or round wells; pyramid, round, or V-bottoms to match your assay and culture needs.
•Automation-friendly fixed plate orientation for liquid handlers.
•Sealing ecosystem — capmats, gas-permeable airporous seals, foil seals, and lids.
•Collection-plate role — used as the receiving plate beneath filter plates in clarification workflows.
• Made in the USA