Learn More
Waters Corp Waters 96-Well Plate, Extended Glass Inserts, 1 mL, Polypropylene
Catalog No. 50780505 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well sample collection plate with 1 mL glass inserts is ideal for autosamplers. It ensures samples only contact glass, preventing adherence to plastic. SBS/ANSI compliant, it also functions as a collection plate for SPE and filtration formats. The polypropylene plate provides chemical resistance, with each well tested for cleanliness and dimensional tolerances.
  • Sample collection plate with 96 wells and 1 mL glass inserts.
  • Designed for autosamplers.
  • Glass inserts ensure samples contact only glass surfaces.
  • SBS/ANSI compliant for robot-compatible systems.
  • Functions as a collection plate for SPE and filtration formats.
  • Polypropylene plate offers chemical resistance.
  • Accommodates compounds that may stick to plastic surfaces via polar glass inserts.
  • Strictly tested for cleanliness and dimensional tolerances.
Catalog No. 50-780-505 Supplier Waters Corp Supplier No. 186000855
This 96-well sample collection plate with 1 mL glass inserts is ideal for autosamplers. It ensures samples only contact glass, preventing adherence to plastic. SBS/ANSI compliant, it also functions as a collection plate for SPE and filtration formats. The polypropylene plate provides chemical resistance, with each well tested for cleanliness and dimensional tolerances.
  • Sample collection plate with 96 wells and 1 mL glass inserts.
  • Designed for autosamplers.
  • Glass inserts ensure samples contact only glass surfaces.
  • SBS/ANSI compliant for robot-compatible systems.
  • Functions as a collection plate for SPE and filtration formats.
  • Polypropylene plate offers chemical resistance.
  • Accommodates compounds that may stick to plastic surfaces via polar glass inserts.
  • Strictly tested for cleanliness and dimensional tolerances.