Learn More
Lonza Walkersville LAL Reagent Grade Multi-well Plates
Catalog No. NC9050345
Encompass
These 96-well plates can be used with the QCL-1000 Endpoint Chromogenic LAL Assay, Kinetic-QCL Kinetic Chromogenic LAL Assay, PYROGENT-5000 Kinetic Turbidimetric LAL Assay and PryoGene rFC Assay. The plates are certified to contain <0.0005 EU/well endotoxin. Each case contains 50 individually wrapped plates.Benefits:Individually wrapped Certified to contain less than 0.0005 EU/wellCertified for compatibility with endotoxin detection assaysCertified to be free from inhibitionApplications:To be used with Kinetic-QCL Kinetic Chromogenic LAL Assay PYROGENT-5000 Kinetic Turbidimetric Assay PyroGene rFC Endpoint Fluorescent Assay QCL-1000 Endpoint Chromogenic LAL Assay.
Catalog No. NC9050345 Supplier Lonza Walkersville Supplier No. 25340
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
These 96-well plates can be used with the QCL-1000 Endpoint Chromogenic LAL Assay, Kinetic-QCL Kinetic Chromogenic LAL Assay, PYROGENT-5000 Kinetic Turbidimetric LAL Assay and PryoGene rFC Assay. The plates are certified to contain <0.0005 EU/well endotoxin. Each case contains 50 individually wrapped plates.Benefits:Individually wrapped Certified to contain less than 0.0005 EU/wellCertified for compatibility with endotoxin detection assaysCertified to be free from inhibitionApplications:To be used with Kinetic-QCL Kinetic Chromogenic LAL Assay PYROGENT-5000 Kinetic Turbidimetric Assay PyroGene rFC Endpoint Fluorescent Assay QCL-1000 Endpoint Chromogenic LAL Assay.