Learn More
Andwin Scientific TPP Multi-well Plates, 96 Wells, Flat, 108/cs
SDP
Catalog No. NC1665139
Encompass
Air vents in the base of the plates allow for consistent air flow between stacked plates. The consistent air flow ensures even temperature distribution, condensation prevention, and greater well to well consistency.
Catalog No. NC1665139 Supplier Andwin Scientific Supplier No. TP92696
Air vents in the base of the plates allow for consistent air flow between stacked plates. The consistent air flow ensures even temperature distribution, condensation prevention, and greater well to well consistency.