Learn More
NEST SCIENTIFIC INC 96 Well Cell Culture Plate, flat, TC, sterile, Bulk, 100/cs
Catalog No. NC2778117
Encompass_Preferred
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. Plates are untreated to support suspended cell lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC2778117 Supplier NEST SCIENTIFIC INC Supplier No. 701002
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. Plates are untreated to support suspended cell lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.