Learn More
Corning™ Half-area 96-well Flat Bottom UV-transparent Microplate without Lid PROMO
Catalog No. 07-200-848S24 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-200-848S24 Clear
1 options
Catalog No. 07-200-848S24 Supplier Corning™ Supplier No. 07200848S24
These 96-well Flat Bottom UV-transparent Microplates have a unique UV-transparent bottom, which is ideal for determining protein and/or nucleic acid concentrations.
  • UV-transparent bottom is molded to the top without adhesives for greater strength and maximum leak resistance
  • For low background and consistent performance at 260 nm and 280 nm
  • Broad linear detection range allows reliable detection of high and low concentrations of biomolecules
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified RNase/DNase-free
Format Half-Area
Lid Without Lid
Array 8 x 12
Color Clear
Volume (Metric) Well 205 μL
No. of Wells 96
Volume (Metric) Working 25 to 125 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 50 Plates/Cs.
No. per Pack 25 Plates/Pk.
Well Color Clear