Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc 96 Well, Flat Bottom, Tissue Culture Plate with Lid, Sterile, 5/Sleeve, 100/CS
SDP

Catalog No. 50147426
Encompass_Preferred

Multiple Well Plate, 96 Well Flat Bottom, Tissue Culture Plate with Lid, 5/Sleeve (not individual pack)
Polystyrene, Tissue Culture Treated for cell attachment.
Single direction lid.
Sterile (gamma irradiated).
5PK/100CS

Plates are labeled with alpha-numeric markings for easy identification of individual wells
Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross-contamination
Lids have a stacking feature for added stability when handling tall stacks of plates

Sterile: Yes
Cell Growth Area: 0.33cm²
Tissue Culture Treated Surface (TCT): Yes
Color / Color Code: Clear
Material: Polystyrene
Shelf Life: 36 months from date of manufacture

Catalog No. 50-147-426 Supplier Quality Biological Inc Supplier No. P886229197
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Multiple Well Plate, 96 Well Flat Bottom, Tissue Culture Plate with Lid, 5/Sleeve (not individual pack)
Polystyrene, Tissue Culture Treated for cell attachment.
Single direction lid.
Sterile (gamma irradiated).
5PK/100CS

Plates are labeled with alpha-numeric markings for easy identification of individual wells
Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross-contamination
Lids have a stacking feature for added stability when handling tall stacks of plates

Sterile: Yes
Cell Growth Area: 0.33cm²
Tissue Culture Treated Surface (TCT): Yes
Color / Color Code: Clear
Material: Polystyrene
Shelf Life: 36 months from date of manufacture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.