Learn More
Stellar Scientific 96-WELL POLYPROPYLENE 2ML WELL
SDP
Catalog No. NC1845175
Encompass
96-Well Polypropylene 2mL Round Well Deep Well Plate, Round Bottom, STERILE, 50/CS
Catalog No. NC1845175 Supplier Stellar Scientific Supplier No. DWP7651S0C
96-Well Polypropylene 2mL Round Well Deep Well Plate, Round Bottom, STERILE, 50/CS