Learn More
Greiner Bio-One 96-well Polypropylene Conical Bottomed Microplates
GREENER_CHOICE
Catalog No. 07000157 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-157 Clear
1 options
Catalog No. 07-000-157 Supplier Greiner Bio-One Supplier No. 651261
Add to Cart
Add to Cart
For high-throughput screening and assay
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Ideally suited for agglutination tests
  • No sharp corners, which means that they can be pipetted easily and cleanly
  • Suitable for +/- analyses
  • Solid bottom
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
Color Clear
Product Type Storage Microplate
Material Polypropylene
Volume (Metric) Well 50 to 335 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 340 μL
Well Shape Round, Conical Bottom (V-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
No. per Pack 10
Quantity 100/Cs.
Well Color Clear
Well Design Chimney