Learn More
Greiner Bio-One 96-well Polypropylene PCR Plate
Catalog No. 07000997 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-997 Clear
1 options
Catalog No. 07-000-997 Supplier Greiner Bio-One Supplier No. 652230P
Add to Cart
Add to Cart
Offers comprehensive thermal cycler compatibility.
  • Suitable for 8-cap strips, adhesive sealers; heat-sealing
  • High-visibility alphanumeric printing for rapid sample identification
  • Packaging labeled with product symbols for easier identification, warehousing
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Color Clear
Product Type PCR Plate
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 0.2 mL
Well Design Rigid style