Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips
GREENER_CHOICE

Catalog No. 07000844 Shop All Greiner Bio-One Products
Change view
Click to view available options
Surface Treatment:
FLUOTRAC 200
FLUOTRAC 600
LUMITRAC 200
LUMITRAC 600
Microlon 200
Microlon 600
For Use With (Application):
Fluorescence measurements
Luminescence measurements
Transmission measurements
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment For Use With (Application)
07-000-844 Microlon 600 Transmission measurements
07-000-843 Microlon 200 Transmission measurements
07-000-845 LUMITRAC 600 Luminescence measurements
07-000-846 LUMITRAC 200 Luminescence measurements
07-000-847 FLUOTRAC 200 Fluorescence measurements
07-000-848 FLUOTRAC 600 Fluorescence measurements
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07-000-844 Supplier Greiner Bio-One Supplier No. 762071
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Treated specifically to increase protein binding. Greiner Bio-One 96 Well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips are suitable for optical measurements.

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Flat Bottom
  • Polystyrene

Service Plan

No

TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Array 8 x 12
Binding Type High Binding
Product Type Strip Plate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 20 to 350 μL
Sterility Non-sterile
Surface Treatment Microlon 600
Total Volume (Metric) Well 388 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Transmission measurements
No. per Case 100
No. per Pack 5
RNase Free Yes
Skirt Color White
Temperature Range (Metric) -20°C to +60°C
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.