Learn More
Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 2 x 8 Strips
GREENER_CHOICE
Catalog No. 07000839 Shop All Greiner Bio-One Products
Change view
Click to view available options
Surface Treatment:
Microlon 200
Microlon 600
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
07-000-839 Microlon 200
07-000-840 Microlon 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-839 Supplier Greiner Bio-One Supplier No. 756070
Add to Cart
Add to Cart
Suitable for optical measurements. Greiner Bio-One 96 Well Polystyrene Strip Plates, 2 x 8 Strips, F-Bottom are suitable for non polar proteins and peptides.
  • Free of detectable DNase, RNase, human DNA
  • 2 x 8 strips
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Suitable for transmission measurements
  • Polystyrene surface is more hydrophobic than surface of hi
TRUSTED_SUSTAINABILITY
Specifications
Format Strip Plate
Binding Type Medium Binding
Color White
Material Polystyrene
No. of Wells 6
Volume (Metric) Working 20 to 350 μL
Sterility Non-sterile
Surface Treatment Microlon 200
Total Volume (Metric) Well 388 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
No. per Case 100
No. per Pack 5
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Well Color Clear