Learn More
Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 2 x 8 Strips
GREENER_CHOICE
Catalog No. 07000840 Shop All Greiner Bio-One Products
Change view
Click to view available options
Surface Treatment:
Medium Binding
Microlon 600
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
07-000-840 Microlon 600
07-000-839 Medium Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-840 Supplier Greiner Bio-One Supplier No. 756071
Add to Cart
Add to Cart
Suitable for optical measurements. Greiner Bio-One 96 Well Polystyrene Strip Plates, 2 x 8 Strips, F-Bottom are suitable for non polar proteins and peptides.
  • Free of detectable DNase, RNase, human DNA
  • 2 x 8 strips
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Suitable for transmission measurements
  • Polystyrene surface
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Strip Plate
Array 8 x 12
Binding Type High Binding
Color Clear
Product Type ELISA Strip Plate
Material Polystyrene
Volume (Metric) Well 388 μL
No. of Wells 96
Volume (Metric) Working 20 to 350 μL
Sterility Non-sterile
Surface Treatment Microlon 600
Total Volume (Metric) Well 388 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
No. per Case 100
No. per Pack 5
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Well Color Clear