Learn More
Analytical Sales & Services 96-Well Polystyrene Microtitration Plate, 250µL V-Bottom, STERILE
Catalog No. 5000804374
Encompass
96-Well Polystyrene Microtitration Plate\n\n• V-Bottom\n• 250µL/Well\n• STERILE\n• Individually Wrapped\n\nQty/Pkg: 50\n\n 
Catalog No. 50-008-04374 Supplier Analytical Sales & Services Supplier No. 960138
96-Well Polystyrene Microtitration Plate\n\n• V-Bottom\n• 250µL/Well\n• STERILE\n• Individually Wrapped\n\nQty/Pkg: 50\n\n 