Learn More
Analytical Sales & Services 96-Well Polystyrene Microtitration Plate, 300µL U-Bottom, STERILE
Catalog No. 5000805812
Encompass
96-Well Polystyrene Microtitration Plate\n\n• U-Bottom\n• 300µL/Well\n• STERILE\n• Individually Wrapped\n\nQty/Pkg: 50
Catalog No. 50-008-05812 Supplier Analytical Sales & Services Supplier No. 960038
96-Well Polystyrene Microtitration Plate\n\n• U-Bottom\n• 300µL/Well\n• STERILE\n• Individually Wrapped\n\nQty/Pkg: 50