Learn More
Arcticwhite LLC 96 Well Reservoir, Pyramid Bottom, Clear / Non-Sterile, Polypropylene (PP), 287 ML, 25 Reagent Reservoirs per Case
SDP
Catalog No. NC9719947
Encompass
96 Well Reservoir, Pyramid Bottom, Clear / Non-Sterile, Polypropylene (PP), 287 ML, 25 Reagent Reservoirs per Case. This 96-well reagent reservoir has a pyramid bottom. It suits automated liquid handlers and multichannel pipetting. Supplied by ArcticWhite as 25 reagent reservoirs per case.
Catalog No. NC9719947 Supplier Arcticwhite LLC Supplier No. AWLSS30014
96 Well Reservoir, Pyramid Bottom, Clear / Non-Sterile, Polypropylene (PP), 287 ML, 25 Reagent Reservoirs per Case. This 96-well reagent reservoir has a pyramid bottom. It suits automated liquid handlers and multichannel pipetting. Supplied by ArcticWhite as 25 reagent reservoirs per case.