Learn More
USA Scientific Inc TempPlate non-skirted polypropylene 0.2 mL 96-well PCR plate, natural, 10/pack
Catalog No. NC9107451
Encompass
TempPlate non-skirted polypropylene 0.2 mL 96-well PCR plate, natural, 10/pack
Catalog No. NC9107451 Supplier USA Scientific Inc Supplier No. 14029596
TempPlate non-skirted polypropylene 0.2 mL 96-well PCR plate, natural, 10/pack